ENA346975289 Enamine

4-(dimethylamino)quinoline-3-carboxylic acid

  • MDL number MFCD11868253
  • Cas Number : 933694-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121...123
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96482

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96482
Availability details for
Quantity
  • Availability
  • Add to Cart