ENA346975290 Enamine

2-methoxy-3,3-dimethylbutan-1-amine

  • MDL number MFCD16065045
  • Cas Number : 1250143-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96483
Availability details for
Quantity
  • Availability
  • Add to Cart