ENA346975303 Enamine

2-amino-N-(2-nitrophenyl)acetamide

  • MDL number MFCD12145784
  • Cas Number : 24715-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96504

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96504
Availability details for
Quantity
  • Availability
  • Add to Cart