ENA346975307 Enamine

2-bromo-4-methylbenzene-1-sulfonamide

  • MDL number MFCD11104196
  • Cas Number : 89694-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96515

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96515
Availability details for
Quantity
  • Availability
  • Add to Cart