ENA346975308 Enamine

3-(3-ethoxy-4-methoxyphenyl)prop-2-enenitrile

  • Cas Number : 757962-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96516
Availability details for
Quantity
  • Availability
  • Add to Cart