ENA346975313 Enamine

3-(3-chlorophenyl)pent-2-enoic acid

  • MDL number MFCD11172818
  • Cas Number : 1049160-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96524

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96524
Availability details for
Quantity
  • Availability
  • Add to Cart