ENA346975315 Enamine

3-(2-bromophenyl)-2-[(2-bromophenyl)methyl]propanoic acid

  • MDL number MFCD20731233
  • Cas Number : 55223-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96531
Availability details for
Quantity
  • Availability
  • Add to Cart