ENA346975321 Enamine

3-(2-aminoethyl)-4,5-dihydro-1H-1,2,4-triazol-5-one hydrochloride

  • MDL number MFCD09864248
  • Cas Number : 879635-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96544
Availability details for
Quantity
  • Availability
  • Add to Cart