ENA346975323 Enamine

3-(5-methyl-1H-indol-3-yl)propanoic acid

  • MDL number MFCD10035276
  • Cas Number : 7394-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96546
Availability details for
Quantity
  • Availability
  • Add to Cart