ENA346975330 Enamine

N,N-dimethyl-2-(phenylamino)acetamide

  • MDL number MFCD01733368
  • Cas Number : 14307-89-2
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-96577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96577
Availability details for
Quantity
  • Availability
  • Add to Cart