ENA346975342 Enamine

3-(2-methylpropyl)pyrrolidine

  • MDL number MFCD07383547
  • Cas Number : 959238-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-96598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96598
Availability details for
Quantity
  • Availability
  • Add to Cart