ENA346975343 Enamine

4-(2-methylpropyl)pyrrolidin-2-one

  • MDL number MFCD09864501
  • Cas Number : 61312-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96599
Availability details for
Quantity
  • Availability
  • Add to Cart