ENA346975344 Enamine

1-[4-(trifluoromethyl)-1,3-thiazol-2-yl]propan-1-amine

  • MDL number MFCD14628825
  • Cas Number : 1248076-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96600
Availability details for
Quantity
  • Availability
  • Add to Cart