ENA346975348 Enamine

(5-methyl-1,3-oxazol-2-yl)methanamine

  • MDL number MFCD09884236
  • Cas Number : 1017228-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-96605

Safety Information

signalword Danger
pcodes P264,P270,P280,P301+P310,P305+P351+P338,P310,P330,P405,P501
hcodes T,Xi
MSDS https://www.enaminestore.com/catalog/EN300-96605
pictograms GHS05,GHS06
hstatements H301,H318
Availability details for
Quantity
  • Availability
  • Add to Cart