ENA346975349 Enamine

cyclooctanecarbaldehyde

  • MDL number MFCD00001747

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-96608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96608
Availability details for
Quantity
  • Availability
  • Add to Cart