ENA346975350 Enamine

2-ethylthiomorpholine

  • MDL number MFCD19219254
  • Cas Number : 30188-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93853
Availability details for
Quantity
  • Availability
  • Add to Cart