ENA346975354 Enamine

ethyl 3-(3,5-dichlorophenyl)-3-hydroxypropanoate

  • MDL number MFCD16756879
  • Cas Number : 1249380-24-2
  • Molecular Weight : 263.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96614
Availability details for
Quantity
  • Availability
  • Add to Cart