ENA346975355 Enamine

2,2-difluoroethan-1-amine hydrochloride

  • MDL number MFCD04038283
  • Cas Number : 79667-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96615

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-96615
pictograms GHS05,GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart