ENA346975358 Enamine

2-(1-phenylpropyl)pyrrolidine

  • MDL number MFCD02663487
  • Cas Number : 383127-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96621
Availability details for
Quantity
  • Availability
  • Add to Cart