ENA346975359 Enamine

1-(6-methoxynaphthalen-2-yl)ethan-1-ol

  • MDL number MFCD01632581
  • Cas Number : 77301-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96624

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-96624
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart