ENA346975366 Enamine

bis(4-(2-methoxyphenyl)piperazine-1-carboximidamide); sulfuric acid

  • MDL number MFCD20731248
  • Cas Number : 1354949-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96631

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96631
Availability details for
Quantity
  • Availability
  • Add to Cart