ENA346975372 Enamine

1-(3-methylphenoxy)-3-[(propan-2-yl)amino]propan-2-ol hydrochloride

  • MDL number MFCD01696861
  • Cas Number : 306-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-96641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96641
Availability details for
Quantity
  • Availability
  • Add to Cart