ENA346975374 Enamine

4-(4-methoxyphenyl)piperidine

  • MDL number MFCD03839927
  • Cas Number : 67259-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-96653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96653
Availability details for
Quantity
  • Availability
  • Add to Cart