ENA346975377 Enamine

5-bromo-1-(4-methylphenyl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD12111880
  • Cas Number : 1156275-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96657

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96657
Availability details for
Quantity
  • Availability
  • Add to Cart