ENA346975378 Enamine

N-(3-aminophenyl)-N-methylacetamide

  • MDL number MFCD02657491
  • Cas Number : 61679-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-117
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-96659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96659
Availability details for
Quantity
  • Availability
  • Add to Cart