ENA346975381 Enamine

4-aminooxane-4-carboxylic acid

  • MDL number MFCD06656112
  • Cas Number : 39124-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96666

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96666
Availability details for
Quantity
  • Availability
  • Add to Cart