ENA346975393 Enamine

3-[(1-methyl-1H-pyrazol-4-yl)methyl]pyrrolidine-2,5-dione

  • MDL number MFCD20326501
  • Cas Number : 1339095-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96682
Availability details for
Quantity
  • Availability
  • Add to Cart