ENA346975401 Enamine

methyl[(1-methyl-1H-1,2,4-triazol-5-yl)methyl]amine

  • MDL number MFCD16851316
  • Cas Number : 1250355-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96693
Availability details for
Quantity
  • Availability
  • Add to Cart