ENA346975402 Enamine

4-phenoxypyridine-2-carboxylic acid

  • MDL number MFCD11181387
  • Cas Number : 99970-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96694

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96694
Availability details for
Quantity
  • Availability
  • Add to Cart