ENA346975412 Enamine

4-chloro-3-(5-cyclopropyl-1H-1,2,3,4-tetrazol-1-yl)aniline

  • MDL number MFCD16676674
  • Cas Number : 1248147-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96714
Availability details for
Quantity
  • Availability
  • Add to Cart