ENA346975416 Enamine

1-[4-(pyrimidin-5-yl)phenyl]ethan-1-amine

  • MDL number MFCD16829946
  • Cas Number : 1250290-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96726
Availability details for
Quantity
  • Availability
  • Add to Cart