ENA346975418 Enamine

2-methoxy-1,3-benzothiazol-6-amine

  • MDL number MFCD18298720
  • Cas Number : 77563-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96728
Availability details for
Quantity
  • Availability
  • Add to Cart