ENA346975419 Enamine

1-(oxan-4-yl)propan-1-amine

  • MDL number MFCD19611850
  • Cas Number : 854697-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96731
Availability details for
Quantity
  • Availability
  • Add to Cart