ENA346975422 Enamine

4-(1,3-dioxaindan-5-yl)-1H-pyrazol-5-amine

  • MDL number MFCD14657238
  • Cas Number : 40545-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94384
Availability details for
Quantity
  • Availability
  • Add to Cart