ENA346975433 Enamine

3-[3-(trifluoromethyl)phenyl]butan-1-amine

  • MDL number MFCD16083222
  • Cas Number : 1225961-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96801
Availability details for
Quantity
  • Availability
  • Add to Cart