ENA346975440 Enamine

N-methyl-2-(piperidin-4-yl)acetamide

  • MDL number MFCD09933326
  • Cas Number : 70724-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96814

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96814
Availability details for
Quantity
  • Availability
  • Add to Cart