ENA346975445 Enamine

4-(1,3-dioxaindan-5-yl)-3-cyclopropyl-1H-pyrazol-5-amine

  • MDL number MFCD11209121
  • Cas Number : 1312617-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94641
Availability details for
Quantity
  • Availability
  • Add to Cart