ENA358812880 Enamine

3-chloro-4-ethoxy-1,1,1-trifluorobut-3-en-2-one

  • MDL number MFCD06660658
  • Cas Number : 119577-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35140

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35140
Availability details for
Quantity
  • Availability
  • Add to Cart