ENA358812884 Enamine

tert-butyl 3-methyl-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine-4-carboxylate

  • MDL number MFCD21334166
  • Cas Number : 1375473-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97873
Availability details for
Quantity
  • Availability
  • Add to Cart