ENA358812890 Enamine

tert-butyl 6-methyl-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine-4-carboxylate

  • MDL number MFCD21334172
  • Cas Number : 1375474-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-97881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97881
Availability details for
Quantity
  • Availability
  • Add to Cart