ENA358812891 Enamine

2,2,2-trifluoro-1-(4-methyl-1,3-thiazol-2-yl)ethan-1-one

  • MDL number MFCD11870461
  • Cas Number : 174824-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248...250
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23115

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-23115
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart