ENA358812897 Enamine

1-(1H-1,3-benzodiazol-2-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD09051201
  • Cas Number : 937629-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37031
Availability details for
Quantity
  • Availability
  • Add to Cart