ENA358812899 Enamine

1-{4-hydroxy-3-[(morpholin-4-yl)methyl]phenyl}ethan-1-one

  • MDL number MFCD06357371
  • Cas Number : 92041-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40683
Availability details for
Quantity
  • Availability
  • Add to Cart