ENA358812900 Enamine

3-(4-fluorophenyl)cyclobutan-1-one

  • MDL number MFCD11848523
  • Cas Number : 143589-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43630

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-43630
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart