ENA358812904 Enamine

3-(3-methylphenyl)cyclobutan-1-one

  • MDL number MFCD11848454
  • Cas Number : 243867-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43634
Availability details for
Quantity
  • Availability
  • Add to Cart