ENA358812905 Enamine

3-(3-fluorophenyl)cyclobutan-1-one

  • MDL number MFCD11848522
  • Cas Number : 1080636-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43635
Availability details for
Quantity
  • Availability
  • Add to Cart