ENA358812907 Enamine

3-(3-bromophenyl)cyclobutan-1-one

  • MDL number MFCD08706340
  • Cas Number : 885267-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43637
Availability details for
Quantity
  • Availability
  • Add to Cart