ENA358812923 Enamine

1-{[1-(difluoromethyl)-1H-imidazol-2-yl]methyl}piperazine

  • MDL number MFCD08444279
  • Cas Number : 923233-19-6
  • Molecular Weight : 216.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57436
Availability details for
Quantity
  • Availability
  • Add to Cart