ENA358812924 Enamine

N-(2-aminoethyl)-3-methoxybenzamide

  • MDL number MFCD04355545
  • Cas Number : 789447-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57538
Availability details for
Quantity
  • Availability
  • Add to Cart