ENA358812942 Enamine

oxetan-2-one

  • MDL number MFCD00005169
  • Cas Number : 57-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 70
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68177

Safety Information

signalword Danger
pcodes P201,P202,P260,P264,P271,P280,P281,P284,P302+P352,P304+P340,P305+P351+P338,P308+P313,P310,P320,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-68177
pictograms GHS06,GHS08
hstatements H315,H319,H330,H350
Availability details for
Quantity
  • Availability
  • Add to Cart